4-(Trifluormethoxy)toluol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

390402-1G
SIAL390402-1GEA 31.3 EUR
SIAL390402-1G
4-(Trifluormethoxy)toluol, Sigma-Aldrich®
4-(Trifluormethoxy)toluol
Formel: C₈H₇F₃O
Molecular Weight: 176,1400 g/mol
MDL: MFCD00042413
CAS-Nummer: 706-27-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR