Methyl-2,2-difluor-2-(fluorsulfonyl)acetat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

390755-5G
SIAL390755-5GEA 32.9 EUR
SIAL390755-5G
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat, Sigma-Aldrich®
Methyl-2,2-difluor-2-(fluorsulfonyl)acetat
Formel: C₃H₃F₃O₄S
Molecular Weight: 192,11 g/mol
MDL: MFCD00144316
CAS-Nummer: 680-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR