3-Azetidincarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Azetanecarboxylic acid, Azetidine-3-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

391131-250MG
SIAL391131-250MGEA 97.4 EUR
SIAL391131-250MG
3-Azetidincarbonsäure, Sigma-Aldrich®
3-Azetidincarbonsäure
Formel: C₄H₇NO₂
Molecular Weight: 101,11 g/mol
MDL: MFCD00191763
CAS-Nummer: 36476-78-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR