1-Acetylpyren, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Pyren-1-yl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

391425-10G
SIAL391425-10GEA 188 EUR
SIAL391425-10G
1-Acetylpyren, Sigma-Aldrich®
1-Acetylpyren
Formel: C₁₈H₁₂O
Molecular Weight: 244,29 g/mol
MDL: MFCD00021663
CAS-Nummer: 3264-21-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR