6-Nitro-m-anissäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Methoxy-2-nitrobenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

391999-25G
SIAL391999-25GEA 59.6 EUR
SIAL391999-25G
6-Nitro-m-anissäure, Sigma-Aldrich®
6-Nitro-m-anissäure
Formel: C₈H₇NO₅
Molecular Weight: 197,15 g/mol
Schmelzpunkt: 138 °C
MDL: MFCD00151836
CAS-Nummer: 1882-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR