Tris(2,4,6-trimethoxyphenyl)phosphin, Sigma-Aldrich®

Lieferant: Merck

392081-5G
SIAL392081-5GEA 200 EUR
SIAL392081-5G
Tris(2,4,6-trimethoxyphenyl)phosphin, Sigma-Aldrich®
Tris(2,4,6-trimethoxyphenyl)phosphin
Formel: C₂₇H₃₃O₉P
Molecular Weight: 532,519 g/mol
MDL: MFCD00145467
CAS-Nummer: 91608-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR