4-Brom-2-fluorbenzoesäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

392383-5G
SIAL392383-5GEA 64.8 EUR
SIAL392383-5G
4-Brom-2-fluorbenzoesäure, Sigma-Aldrich®
4-Bromo-2-fluorobenzoic acid
Formel: C₇H₄BrFO₂
Molecular Weight: 219,01 g/mol
MDL: MFCD00143263
CAS-Nummer: 112704-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR