Diethyl(hydroxymethyl)phosphonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

392626-25ML
SIAL392626-25MLEA 241 EUR
SIAL392626-25ML
Diethyl(hydroxymethyl)phosphonat, Sigma-Aldrich®
Diethyl(hydroxymethyl)phosphonat
Formel: C₅H₁₃O₄P
Molecular Weight: 168,13 g/mol
MDL: MFCD00014418
CAS-Nummer: 3084-40-0
EINECS: 221-391-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR