4,5-Dimethoxy-2-nitrobenzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Nitroveratrumylalkohol, 6-Nitroveratrylalkohol, (4,5-Dimethoxy-2-nitrophenyl)methanol

392847-10G
SIAL392847-10GEA 359 EUR
SIAL392847-10G
4,5-Dimethoxy-2-nitrobenzylalkohol, Sigma-Aldrich®
4,5-Dimethoxy-2-nitrobenzylalkohol
Formel: C₉H₁₁NO₅
Molecular Weight: 213,19 g/mol
MDL: MFCD00014701
CAS-Nummer: 1016-58-6
EINECS: 213-806-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR