15-Hydroxypentadecansäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

392979-5G
SIAL392979-5GEA 315 EUR
SIAL392979-5G
15-Hydroxypentadecansäure, Sigma-Aldrich®
15-Hydroxypentadecanoic acid
Formel: C₁₅H₃₀O₃
MDL: MFCD00039535
CAS-Nummer: 4617-33-8
EINECS: 225-026-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR