4'-Aminobenzo-15-krone-5 (4'-Aminobenzo-1,4,7,10,13-pentaoxacyclopentadecan), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3-(4-Aminobenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene, 4-Aminobenzo-15-crown 5-Ether, 4'-Aminobenzo-15-crown 5-Ether

M-Clarity™ Qualitätsstufe = MQ100

393428-500MG
SIAL393428-500MGEA 134 EUR
SIAL393428-500MG
4'-Aminobenzo-15-krone-5 (4'-Aminobenzo-1,4,7,10,13-pentaoxacyclopentadecan), Sigma-Aldrich®
4'-Aminobenzo-15-krone-5 (4'-Aminobenzo-1,4,7,10,13-pentaoxacyclopentadecan)
Formel: C₁₄H₂₁NO₅
Molecular Weight: 283.3202 g/mol
MDL: MFCD00068686
CAS-Nummer: 60835-71-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR