3-(2-Hydroxyphenyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(2-Hydroxyphenyl)propansäure

393533-10G
SIAL393533-10GEA 46.1 EUR
SIAL393533-10G
3-(2-Hydroxyphenyl)propionsäure, Sigma-Aldrich®
3-(2-Hydroxyphenyl)propionsäure
Formel: C₉H₁₀O₃
MDL: MFCD00014026
CAS-Nummer: 495-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR