3-Chlor-2-fluor-5-(trifluormethyl)pyridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

393576-5G
SIAL393576-5GEA 87.1 EUR
SIAL393576-5G
3-Chlor-2-fluor-5-(trifluormethyl)pyridin, Sigma-Aldrich®
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₄N
Molecular Weight: 199,53 g/mol
MDL: MFCD00191918
CAS-Nummer: 72537-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR