2-(Hydroxymethyl)-1,3-propandiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tris(hydroxymethyl)methane, 2-(Hydroxymethyl)propane-1,3-diol

M-Clarity™ Qualitätsstufe = MQ100

393657-1G
SIAL393657-1GEA 163 EUR
SIAL393657-1G
2-(Hydroxymethyl)-1,3-propandiol, Sigma-Aldrich®
2-(Hydroxymethyl)-1,3-propanediol
Formel: C₄H₁₀O₃
Molecular Weight: 106,12 g/mol
MDL: MFCD00192217
CAS-Nummer: 4704-94-3
EINECS: 225-187-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR