2-Hydroxy-5-methylisophthalaldehyd, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-5-methylbenzene-1,3-dicarbaldehyde

M-Clarity™ Qualitätsstufe = MQ100

393851-1G
SIAL393851-1GEA 72.7 EUR
SIAL393851-1G
2-Hydroxy-5-methylisophthalaldehyd, Sigma-Aldrich®
2-Hydroxy-5-methylisophthalaldehyd
Formel: C₉H₈O₃
Molecular Weight: 164,1600 g/mol
MDL: MFCD00016596
CAS-Nummer: 7310-95-4
EINECS: 230-768-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR