2-Ethylphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2-Ethylphenyl)hydrazin Hydrochlorid, 2-Ethylphenylhydrazin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

393916-10G
SIAL393916-10GEA 113 EUR
SIAL393916-10G
2-Ethylphenylhydraziniumchlorid, Sigma-Aldrich®
2-Ethylphenylhydraziniumchlorid
Formel: C₈H₁₃ClN₂
Molecular Weight: 172,66 g/mol
MDL: MFCD00071599
CAS-Nummer: 19398-06-2
EINECS: 421-460-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR