2-Amino-4-phenylphenol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Amino-4-hydroxybiphenyl, 3-Amino-[1,1'-biphenyl]-4-ol

M-Clarity™ Qualitätsstufe = MQ100

394114-1G
SIAL394114-1GEA 57 EUR
SIAL394114-1G
2-Amino-4-phenylphenol, Sigma-Aldrich®
2-Amino-4-phenylphenol
Formel: C₁₂H₁₁NO
Molecular Weight: 185,2200 g/mol
MDL: MFCD00059187
CAS-Nummer: 1134-36-7
EINECS: 214-484-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR