2-(2-Bromphenyl)ethanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

394548-5G
SIAL394548-5GEA 25.5 EUR
SIAL394548-5G
2-(2-Bromphenyl)ethanol, Sigma-Aldrich®
2-(2-Bromphenyl)ethanol
Formel: C₈H₉BrO
Molecular Weight: 201,06 g/mol
MDL: MFCD00093566
CAS-Nummer: 1074-16-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR