N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ200
394882-100ML
394882-10X1ML
394882-25ML
394882-5ML
SIAL394882-100MLEA
1110
EUR
SIAL394882-100ML
SIAL394882-10X1ML
SIAL394882-25ML
SIAL394882-5ML
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA), Sigma-Aldrich®
N-(tert-Butyldimethylsilyl)-N-methyltrifluoracetamid (MTBSTFA)
Formel:
C₉H₁₈F₃NOSi Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009671 CAS-Nummer: 77377-52-7 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...