Tris(4-fluorphenyl)phosphin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

395099-5G
SIAL395099-5GEA 134 EUR
SIAL395099-5G
Tris(4-fluorphenyl)phosphin, Sigma-Aldrich®
Tris(4-fluorphenyl)phosphin
Formel: C₁₈H₁₂F₃P
Molecular Weight: 316,26 g/mol
MDL: MFCD00013553
CAS-Nummer: 18437-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR