Methyl-3-indolcarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

395307-25G
SIAL395307-25GEA 67.9 EUR
SIAL395307-25G
Methyl-3-indolcarboxylat, Sigma-Aldrich®
Methyl-3-indolcarboxylat
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
MDL: MFCD00189407
CAS-Nummer: 942-24-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR