Tetrachlorbis(tetrahydrofuran)titan(IV), Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
395404-25G
395404-5G
SIAL395404-25GEA
243
EUR
SIAL395404-25G
SIAL395404-5G
Tetrachlorbis(tetrahydrofuran)titan(IV), Sigma-Aldrich®
Tetrachlorbis(tetrahydrofuran)titan(IV)
Formel:
C₄H₈Cl₄OTi |
MDL:
MFCD00077884 CAS-Nummer: 31011-57-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...