4'-(Methylthio)acetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-(Methylthio)phenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

396583-1G
SIAL396583-1GEA 22.5 EUR
SIAL396583-1G
4'-(Methylthio)acetophenon, Sigma-Aldrich®
4'-(Methylthio)acetophenon
Formel: C₉H₁₀OS
MDL: MFCD00039835
CAS-Nummer: 1778-09-2
EINECS: 217-213-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR