Gold(I)iodid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

398411-1G
SIAL398411-1GEA 191 EUR
SIAL398411-1G
Gold(I)iodid, Sigma-Aldrich®
Gold(I)iodid
Formel: AuI
MDL: MFCD00014175
CAS-Nummer: 10294-31-2
EINECS: 233-656-3
UN: 3260
ADR: 8,III
Merck Index: 12,04530

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR