Iridium(IV)oxid Dihydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Iridiumdioxid Dihydrat

M-Clarity™ Qualitätsstufe = MQ100

398500-1G
SIAL398500-1GEA 755 EUR
SIAL398500-1G
Iridium(IV)oxid Dihydrat, Sigma-Aldrich®
Iridium(IV)oxid Dihydrat
Formel: IrO₂·2H₂O
Molecular Weight: 260,25 g/mol
MDL: MFCD00011065
CAS-Nummer: 30980-84-8
EINECS: 234-743-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR