Benzoin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-2-phenylacetophenone

M-Clarity™ Qualitätsstufe = MQ100

399396-5G
SIAL399396-5GEA 103 EUR
SIAL399396-5G
Benzoin, Sigma-Aldrich®
Benzoin
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
MDL: MFCD00004496
CAS-Nummer: 119-53-9
EINECS: 204-331-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR