2,3-Dicyanpyrazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyrazine-2,3-dicarbonitrile

M-Clarity™ Qualitätsstufe = MQ100

399620-5G
SIAL399620-5GEA 106 EUR
SIAL399620-5G
2,3-Dicyanpyrazin, Sigma-Aldrich®
2,3-Dicyanopyrazine
Formel: C₆H₂N₄
Molecular Weight: 130,1069 g/mol
MDL: MFCD00059771
CAS-Nummer: 13481-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR