4,5-Difluoranthranilsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Amino-4,5-difluorobenzoic acid

M-Clarity™ Qualitätsstufe = MQ100

399760-5G
SIAL399760-5GEA 58 EUR
SIAL399760-5G
4,5-Difluoranthranilsäure, Sigma-Aldrich®
4,5-Difluoranthranilsäure
Formel: C₇H₅F₂NO₂
Molecular Weight: 173,12 g/mol
MDL: MFCD00077509
CAS-Nummer: 83506-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR