Titan(III)fluorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
399817-5G
SIAL399817-5GEA
62.5
EUR
SIAL399817-5G
Titan(III)fluorid, Sigma-Aldrich®
Titan(III)fluorid
Formel:
F₃Ti Dichte: 2,98 g/cm³ (20 °C) |
MDL:
MFCD00042544 CAS-Nummer: 13470-08-1 EINECS: 236-732-4 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...