Hafnium(IV)fluorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

399868-25G
SIAL399868-25GEA 0 EUR
SIAL399868-25G
Hafnium(IV)fluorid, Sigma-Aldrich®
Hafnium(IV)fluorid
Formel: F₄Hf
Molecular Weight: 254,48 g/mol
MDL: MFCD00016127
CAS-Nummer: 13709-52-9
EINECS: 237-258-0
UN: 3290
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR