trans-4-Brom-β-nitrostyrol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (E)-1-Bromo-4-(2-nitrovinyl)benzene

M-Clarity™ Qualitätsstufe = MQ200

400017-5G
SIAL400017-5GEA 148 EUR
SIAL400017-5G
trans-4-Brom-β-nitrostyrol, Sigma-Aldrich®
trans-4-Brom-β-nitrostyrol
Formel: C₈H₆BrNO₂
MDL: MFCD00191856
CAS-Nummer: 5153-71-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR