Lithiumtitanat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Lithiumtitanat, Lithiumorthotitanat, Lithiumtitanat(IV)

M-Clarity™ Qualitätsstufe = MQ100

400939-100G
SIAL400939-100GEA 73.6 EUR
SIAL400939-100G
Lithiumtitanat, Sigma-Aldrich®
Lithiumtitanat
Formel: Li₂O₃Ti
Molecular Weight: 109.76 g/mol
MDL: MFCD00016181
CAS-Nummer: 12031-82-2
EINECS: 234-759-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR