Cobalt(II)oxalat Dihydrat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Cobalt(II) oxalate dihydrate

M-Clarity™ Qualitätsstufe = MQ100

401285-50G
SIAL401285-50GEA 253 EUR
SIAL401285-50G
Cobalt(II)oxalat Dihydrat, Sigma-Aldrich®
Cobalt(II)oxalat Dihydrat
Formel: C₂CoO₄·2H₂O
MDL: MFCD03414148
CAS-Nummer: 5965-38-8
EINECS: 212-409-3
UN: 3288
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR