Cobalt(II)perchlorat Hexahydrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

401404-25G
SIAL401404-25GEA 55.4 EUR
SIAL401404-25G
Cobalt(II)perchlorat Hexahydrat, Sigma-Aldrich®
Cobalt(II)perchlorat Hexahydrat
Formel: Cl₂CoO₈·6H₂O
MDL: MFCD00150207
CAS-Nummer: 13478-33-6
EINECS: 236-653-5
UN: 1481
ADR: 5.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR