Calciumperchlorat Tetrahydrat, Sigma-Aldrich®

Lieferant: Merck

401420-100G
SIAL401420-100GEA 59 EUR
SIAL401420-100G
Calciumperchlorat Tetrahydrat, Sigma-Aldrich®
Calciumperchlorat Tetrahydrat
Formel: CaClO₄·4H₂O
MDL: MFCD00191895
CAS-Nummer: 15627-86-8
EINECS: 236-768-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR