Blei(II)acetylacetonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Blei(II) 2,4-pentanedionat
M-Clarity™ Qualitätsstufe = MQ100
401684-5G
SIAL401684-5GEA
78.1
EUR
SIAL401684-5G
Blei(II)acetylacetonat, Sigma-Aldrich®
Blei(II)acetylacetonat
Formel:
C₁₀H₁₄O₄Pb Molecular Weight: 405,41 g/mol |
MDL:
MFCD00013499 CAS-Nummer: 15282-88-9 EINECS: 239-323-9 UN: 3467 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...