Cobalt(2+)bis(1,2,3,4,5-pentamethyl-2,4-cyclopentadienid), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

401781-1G
SIAL401781-1GEA 122 EUR
SIAL401781-1G
Cobalt(2+)bis(1,2,3,4,5-pentamethyl-2,4-cyclopentadienid), Sigma-Aldrich®
Bis(pentamethylcyclopentadienyl)cobalt(II)
Formel: C₂₀H₃₀Co
CAS-Nummer: 74507-62-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR