Methyltetrahydropyran-4-carboxylat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Methyl tetrahydro-2H-pyran-4-carboxylate
40199-50ML
SIAL40199-50MLEA
118
EUR
SIAL40199-50ML
Methyltetrahydropyran-4-carboxylat, Sigma-Aldrich®
Methyltetrahydropyran-4-carboxylat
Formel:
C₇H₁₂O₃ Molecular Weight: 144.1684 g/mol |
MDL:
MFCD01075170 CAS-Nummer: 110238-91-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...