6-Brom-3-formylchromon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Bromo-4-oxo-4H-chromene-3-carbaldehyde

M-Clarity™ Qualitätsstufe = MQ100

402168-1G
SIAL402168-1GEA 31.4 EUR
SIAL402168-1G
6-Brom-3-formylchromon, Sigma-Aldrich®
6-Brom-3-formylchromon
Formel: C₁₀H₅BrO₃
Molecular Weight: 253,05 g/mol
MDL: MFCD00191849
CAS-Nummer: 52817-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR