6-Chlor-3-formyl-7-methylchromon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

402184-1G
SIAL402184-1GEA 31.4 EUR
SIAL402184-1G
6-Chlor-3-formyl-7-methylchromon, Sigma-Aldrich®
6-Chlor-3-formyl-7-methylchromon
Formel: C₁₁H₇ClO₃
MDL: MFCD00191919
CAS-Nummer: 64481-12-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR