(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-N,N-Dimethyl-1-ferrocenylethylamine

40241-1ML
SIAL40241-1MLEA 371 EUR
SIAL40241-1ML
(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamin, Sigma-Aldrich®
(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamin
Formel: C₁₄H₁₉FeN
Molecular Weight: 257,16 g/mol
MDL: MFCD00066272
CAS-Nummer: 31886-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR