N,N-Dimethylformamiddibutylacetal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1-Dibutoxytrimethylamin

40262-10ML
SIAL40262-10MLEA 85.6 EUR
SIAL40262-10ML
N,N-Dimethylformamiddibutylacetal, Sigma-Aldrich®
N,N-Dimethylformamiddibutylacetal
Formel: C₁₁H₂₅NO₂
Molecular Weight: 203,32 g/mol
Siedepunkt: 50 °C (0,5 mmHg)
Dichte: 0,852 g/cm³ (20 °C)
MDL: MFCD00015249
CAS-Nummer: 18503-90-7
EINECS: 242-387-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR