2,2,2,3'-Tetrafluoracetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2,2-Trifluoro-1-(3-fluorophenyl)ethanone

M-Clarity™ Qualitätsstufe = MQ100

402702-10G
SIAL402702-10GEA 194 EUR
SIAL402702-10G
2,2,2,3'-Tetrafluoracetophenon, Sigma-Aldrich®
2,2,2,3'-Tetrafluoracetophenon
Formel: C₈H₄F₄O
Molecular Weight: 192,11 g/mol
MDL: MFCD00039236
CAS-Nummer: 708-64-5
EINECS: 211-903-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR