Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')nickel(II), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), Ni(TMHD)2
M-Clarity™ Qualitätsstufe = MQ100
403393-1G
SIAL403393-1GEA
77.4
EUR
SIAL403393-1G
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')nickel(II), Sigma-Aldrich®
Bis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')nickel(II)
Formel:
C₂₂H₃₈NiO₄ Molecular Weight: 425,23 g/mol |
MDL:
MFCD00192348 CAS-Nummer: 14481-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...