Bortribromid 1 mol/l in Heptan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

403857-100ML
SIAL403857-100MLEA 153 EUR
SIAL403857-100ML
Bortribromid 1 mol/l in Heptan, Sigma-Aldrich®
Bortribromid
Formel: BBr₃
Molecular Weight: 250.52 g/mol
Siedepunkt: 91,2
Schmelzpunkt: -46
MDL: MFCD00011312
CAS-Nummer: 10294-33-4
EINECS: 233-657-9
Merck Index: 13,01337

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR