(Trimethylphosphin)gold(I)chlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Chlorotrimethylphosphinegold(I), Chlorotrimethylphosphine gold(I)

M-Clarity™ Qualitätsstufe = MQ100

404217-1G 404217-250MG
SIAL404217-1GEA 555 EUR
SIAL404217-1G SIAL404217-250MG
(Trimethylphosphin)gold(I)chlorid, Sigma-Aldrich®
(Trimethylphosphine)gold(I) chloride
Formel: C₃H₉AuClP
Molecular Weight: 308.5 g/mol
MDL: MFCD00058861
CAS-Nummer: 15278-97-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR