(Trimethylphosphin)gold(I)chlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Chlorotrimethylphosphinegold(I), Chlorotrimethylphosphine gold(I)
M-Clarity™ Qualitätsstufe = MQ100
404217-1G
404217-250MG
SIAL404217-1GEA
555
EUR
SIAL404217-1G
SIAL404217-250MG
(Trimethylphosphin)gold(I)chlorid, Sigma-Aldrich®
(Trimethylphosphine)gold(I) chloride
Formel:
C₃H₉AuClP Molecular Weight: 308.5 g/mol |
MDL:
MFCD00058861 CAS-Nummer: 15278-97-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...