Benzyl-phenylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzyl Phenyl Ether

404284-5G
SIAL404284-5GEA 28 EUR
SIAL404284-5G
Benzyl-phenylether, Sigma-Aldrich®
Benzyl-phenylether
Formel: C₁₃H₁₂O
Molecular Weight: 184,24 g/mol
MDL: MFCD00020660
CAS-Nummer: 946-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR