N,N'-Bis(3-aminopropyl)propan-1,3-diamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N,N'-Bis(3-aminopropyl)-1,3-propanediamine, 1,5,9,13-Tetraazatridecane

M-Clarity™ Qualitätsstufe = MQ100

404810-5G
SIAL404810-5GEA 271 EUR
SIAL404810-5G
N,N'-Bis(3-aminopropyl)propan-1,3-diamin, Sigma-Aldrich®
N,N'-Bis(3-aminopropyl)propan-1,3-diamin
Formel: C₉H₂₄N₄
Molecular Weight: 188,31 g/mol
MDL: MFCD00008213
CAS-Nummer: 4605-14-5
EINECS: 225-007-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR