1-Methyl-2-phenylindol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

404888-10G
SIAL404888-10GEA 43.8 EUR
SIAL404888-10G
1-Methyl-2-phenylindol, Sigma-Aldrich®
1-Methyl-2-phenylindole
Formel: C₁₅H₁₃N
Molecular Weight: 207,27 g/mol
MDL: MFCD00022892
CAS-Nummer: 3558-24-5
EINECS: 222-618-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR